Learn More
Medchemexpress LLC Nelonicline | 1026134-63-3 | 99.8% | 313.42 g/mol | C17H19N3OS | 10 MG
SDP
Catalog No. 5000288926
Encompass_Preferred
Nelonicline is an orally active, selective agonist of the α7 nicotinic acetylcholine receptor supplied as a research reagent for preclinical and in vitro studies. It has molecular formula C17H19N3OS and molecular weight 313.42 g/mol. For research use only; not for clinical or diagnostic applications.
  • Selective α7 nicotinic acetylcholine receptor agonist suitable for receptor pharmacology studies.
  • Orally active small molecule compatible with in vivo and in vitro experiments.
  • Available as a 10 MG solid and as a 10 mM solution in DMSO for assay use.
  • High-purity research compound appropriate for preclinical studies.
  • Supplied for research use only, not intended for therapeutic use.
Catalog No. 50-002-88926 Supplier Medchemexpress LLC Supplier No. HY1674810MG
Nelonicline is an orally active, selective agonist of the α7 nicotinic acetylcholine receptor supplied as a research reagent for preclinical and in vitro studies. It has molecular formula C17H19N3OS and molecular weight 313.42 g/mol. For research use only; not for clinical or diagnostic applications.
  • Selective α7 nicotinic acetylcholine receptor agonist suitable for receptor pharmacology studies.
  • Orally active small molecule compatible with in vivo and in vitro experiments.
  • Available as a 10 MG solid and as a 10 mM solution in DMSO for assay use.
  • High-purity research compound appropriate for preclinical studies.
  • Supplied for research use only, not intended for therapeutic use.