Learn More
Medchemexpress LLC Nelonicline (ABT-126) | 1026134-63-3 | 99.8% | 313.42 | 100 MG
SDP
Catalog No. 5000393542
Encompass_Preferred
Nelonicline (ABT-126) is an orally active and selective α7 nicotinic receptor agonist with high affinity to α7 nAChRs in human brain (Ki=12.3 nM). It is utilized for the research of schizophrenia and Alzheimer's disease.
  • Orally active and selective α7 nicotinic receptor agonist
  • High affinity to α7 nAChRs in human brain
  • Activates currents in Xenopus oocytes expressing recombinant human α7 nAChRs
  • Used for research of schizophrenia and Alzheimer's disease
  • Binds to α3β4* nAChRs in human IMR-32 neuroblastoma cells
  • Acts as a 5-HT3 receptor antagonist
Catalog No. 50-003-93542 Supplier Medchemexpress LLC Supplier No. HY16748100MG
Nelonicline (ABT-126) is an orally active and selective α7 nicotinic receptor agonist with high affinity to α7 nAChRs in human brain (Ki=12.3 nM). It is utilized for the research of schizophrenia and Alzheimer's disease.
  • Orally active and selective α7 nicotinic receptor agonist
  • High affinity to α7 nAChRs in human brain
  • Activates currents in Xenopus oocytes expressing recombinant human α7 nAChRs
  • Used for research of schizophrenia and Alzheimer's disease
  • Binds to α3β4* nAChRs in human IMR-32 neuroblastoma cells
  • Acts as a 5-HT3 receptor antagonist