Learn More
Medchemexpress LLC HY-10559 5mg Medchemexpress, Nelotanserin CAS:839713-36-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105595MG
Encompass_Preferred
Medchemexpress, HY-10559 5mg Nelotanserin CAS:839713-36-9 Nelotanserin is a potent 5-HT2A inverse agonist, a moderately potent 5-HT2C partial inverse agonist and a weak 5-HT2B inverse agonist, with IC50s of 1.7, 79, 791 nM in IP accumulation assays, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8278