Learn More
Teknova Nematode Growth Media Agar Plates (NGM). 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Supplier:  Teknova N10984PK
Encompass
0.30% Sodium Chloride, 0.25% Tryptone, 2.00% Agar, 5g/mL Cholesterol, 1mM Calcium Chloride, 1mM Magnesium Sulfate, 25mM Potassium Phosphate Buffer pH 6.0
Catalog No. 50-238-7855