Learn More
Medchemexpress LLC HY-19535A 5mg Medchemexpress, Nemiralisib CAS:1254036-71-9 Purity:>98%
SDP
Catalog No. 502082139
Encompass_Preferred
Medchemexpress, HY-19535A 5mg Nemiralisib CAS:1254036-71-9 Nemiralisib (GSK2269557 free base) is a potent and highly selective PI3Kδ inhibitor with a pKi of 9.9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2139 Supplier Medchemexpress LLC Supplier No. HY19535A5MG
Medchemexpress, HY-19535A 5mg Nemiralisib CAS:1254036-71-9 Nemiralisib (GSK2269557 free base) is a potent and highly selective PI3Kδ inhibitor with a pKi of 9.9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.