Learn More
Medchemexpress LLC Nemonoxacin-d3 | 99.9% | 374.45 | 1 MG
SDP
Catalog No. 5000359450
Encompass_Preferred
Nemonoxacin-d3 is a deuterated compound, chemically known as 3-Quinolinecarboxylic acid, 7-[(3S,5S)-3-amino-5-methyl-1-piperidinyl]-1-cyclopropyl-1,4-dihydro-8-methoxy-4-oxo-d3. This compound has a molecular weight of 374.45 and is supplied as a high-purity material, primarily intended for research applications.
  • High purity of 99.9%
  • Confirmed structure by 1H NMR and LCMS
  • Isotopic enrichment of 99.76%
  • Off-white to light yellow solid appearance
  • Stable for extended periods when stored properly
Catalog No. 50-003-59450 Supplier Medchemexpress LLC Supplier No. HY14956S1MG
Nemonoxacin-d3 is a deuterated compound, chemically known as 3-Quinolinecarboxylic acid, 7-[(3S,5S)-3-amino-5-methyl-1-piperidinyl]-1-cyclopropyl-1,4-dihydro-8-methoxy-4-oxo-d3. This compound has a molecular weight of 374.45 and is supplied as a high-purity material, primarily intended for research applications.
  • High purity of 99.9%
  • Confirmed structure by 1H NMR and LCMS
  • Isotopic enrichment of 99.76%
  • Off-white to light yellow solid appearance
  • Stable for extended periods when stored properly