Learn More
Medchemexpress LLC Nemonoxacin malate | 951163-60-3 | 99.29% | 505.52 | 100 MG
SDP
Catalog No. 5000343351
Encompass_Preferred
Nemonoxacin malate is a nonfluorinated quinolone antibiotic with broad-spectrum activity against Gram-positive, Gram-negative, and atypical pathogens. It can inhibit drug-resistant *Streptococcus pneumoniae* and Methicillin-resistant *Staphylococcus aureus* and can be used for the research of community-acquired pneumonia.
  • Nonfluorinated quinolone antibiotic.
  • Broad-spectrum activity against Gram-positive, Gram-negative and atypical pathogens.
  • Inhibits drug-resistant *Streptococcus pneumoniae* and Methicillin-resistant *Staphylococcus aureus*.
  • Used for the research of community-acquired pneumonia.
  • Antibacterial activity against *Chlamydia pneumoniae* with MIC90s of 0.06 μg/mL.
  • Highly active against community-acquired MRSA (CA-MRSA) (MIC90: 0.5 and 0.06 μg/ml), and exerts limited activity against Ciprofloxacin-resistant MRSA (MIC90: 1 μg/ml), and Vancomycin-intermediate MRSA (MIC90: 2 μg/ml).
Catalog No. 50-003-43351 Supplier Medchemexpress LLC Supplier No. HY111023100MG
Nemonoxacin malate is a nonfluorinated quinolone antibiotic with broad-spectrum activity against Gram-positive, Gram-negative, and atypical pathogens. It can inhibit drug-resistant *Streptococcus pneumoniae* and Methicillin-resistant *Staphylococcus aureus* and can be used for the research of community-acquired pneumonia.
  • Nonfluorinated quinolone antibiotic.
  • Broad-spectrum activity against Gram-positive, Gram-negative and atypical pathogens.
  • Inhibits drug-resistant *Streptococcus pneumoniae* and Methicillin-resistant *Staphylococcus aureus*.
  • Used for the research of community-acquired pneumonia.
  • Antibacterial activity against *Chlamydia pneumoniae* with MIC90s of 0.06 μg/mL.
  • Highly active against community-acquired MRSA (CA-MRSA) (MIC90: 0.5 and 0.06 μg/ml), and exerts limited activity against Ciprofloxacin-resistant MRSA (MIC90: 1 μg/ml), and Vancomycin-intermediate MRSA (MIC90: 2 μg/ml).