Learn More
Medchemexpress LLC Nemonoxacin malate | 951163-60-3 | 99.3% | 505.52 | 10 MG
Supplier: Medchemexpress LLC HY11102310MG
Nemonoxacin malate is a nonfluorinated quinolone antibiotic with broad-spectrum activity against Gram-positive, Gram-negative, and atypical pathogens. It can inhibit drug-resistant *Streptococcus pneumoniae* and Methicillin-resistant *Staphylococcus aureus*, and is used for research of community-acquired pneumonia. In vitro, it shows antibacterial activity against *Chlamydia pneumoniae* with MIC90s of 0.06 μg/mL, and is highly active against community-acquired MRSA (CA-MRSA).
- Nonfluorinated quinolone antibiotic
- Broad-spectrum activity against Gram-positive, Gram-negative, and atypical pathogens
- Inhibits drug-resistant *Streptococcus pneumoniae*
- Inhibits Methicillin-resistant *Staphylococcus aureus*
- Used for research of community-acquired pneumonia
- Antibacterial activity against *Chlamydia pneumoniae*
- Highly active against community-acquired MRSA (CA-MRSA)