Learn More
Medchemexpress LLC Nemonoxacin malate | 951163-60-3 | 99.3% | 505.52 | 100 MG
SDP
Catalog No. 5000343183
Encompass_Preferred
Nemonoxacin malate is a nonfluorinated quinolone antibiotic with broad-spectrum activity against various pathogens, including drug-resistant Streptococcus pneumoniae and Methicillin-resistant Staphylococcus aureus. It is used in research for community-acquired pneumonia.
  • Nonfluorinated quinolone antibiotic
  • Broad-spectrum activity against gram-positive, gram-negative, and atypical pathogens
  • Inhibits drug-resistant *Streptococcus pneumoniae*
  • Inhibits methicillin-resistant *Staphylococcus aureus*
  • Used for research on community-acquired pneumonia
Catalog No. 50-003-43183 Supplier Medchemexpress LLC Supplier No. HY11102350MG
Nemonoxacin malate is a nonfluorinated quinolone antibiotic with broad-spectrum activity against various pathogens, including drug-resistant Streptococcus pneumoniae and Methicillin-resistant Staphylococcus aureus. It is used in research for community-acquired pneumonia.
  • Nonfluorinated quinolone antibiotic
  • Broad-spectrum activity against gram-positive, gram-negative, and atypical pathogens
  • Inhibits drug-resistant *Streptococcus pneumoniae*
  • Inhibits methicillin-resistant *Staphylococcus aureus*
  • Used for research on community-acquired pneumonia