Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nemonoxacin malate | 951163-60-3 | MFCD11865196 | 98.3% | 505.52 | C24H31N3O9 | 5 MG
SDP

Catalog No. 5000263345
Encompass_Preferred

Nemonoxacin malate is a nonfluorinated quinolone antibiotic research compound (CAS 951163-60-3) with broad-spectrum activity against Gram-positive, Gram-negative, and atypical pathogens. It has demonstrated activity versus drug-resistant Streptococcus pneumoniae and methicillin-resistant Staphylococcus aureus and is used in studies of community-acquired pneumonia.

  • High purity (98.3%).
  • Molecular weight 505.52 g/mol.
  • Molecular formula C24H31N3O9.
  • Available in small research quantities (e.g., 5 mg).
  • Suitable for in vitro and bacterial infection research.

Catalog No. 50-002-63345 Supplier Medchemexpress LLC Supplier No. HY1110235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nemonoxacin malate is a nonfluorinated quinolone antibiotic research compound (CAS 951163-60-3) with broad-spectrum activity against Gram-positive, Gram-negative, and atypical pathogens. It has demonstrated activity versus drug-resistant Streptococcus pneumoniae and methicillin-resistant Staphylococcus aureus and is used in studies of community-acquired pneumonia.

  • High purity (98.3%).
  • Molecular weight 505.52 g/mol.
  • Molecular formula C24H31N3O9.
  • Available in small research quantities (e.g., 5 mg).
  • Suitable for in vitro and bacterial infection research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.