Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neo 376 | 496921-73-4 | 99.2% | 357.88 | C20H24ClN3O | 5 MG
SDP

Catalog No. 5000248070
Encompass_Preferred

NEO 376 is a small-molecule research compound described as a selective modulator of 5-HT1 (serotonin), GABA, and dopamine receptors with reported antipsychotic activity. It is intended for pharmacological and neurobiological studies and is provided with analytical documentation to support research use.

  • Selective modulator of 5-HT1, GABA, and dopamine receptors.
  • Reported antipsychotic activity suitable for psychiatric research.
  • High purity: 99.2%.
  • Molecular weight 357.88 and formula C20H24ClN3O.
  • Soluble in DMSO; available as 10 mM solution or as solid (small pack sizes).
  • Supplied with datasheet, certificate of analysis, and safety data sheet for research use.

Catalog No. 50-002-48070 Supplier Medchemexpress LLC Supplier No. HY1015835MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NEO 376 is a small-molecule research compound described as a selective modulator of 5-HT1 (serotonin), GABA, and dopamine receptors with reported antipsychotic activity. It is intended for pharmacological and neurobiological studies and is provided with analytical documentation to support research use.

  • Selective modulator of 5-HT1, GABA, and dopamine receptors.
  • Reported antipsychotic activity suitable for psychiatric research.
  • High purity: 99.2%.
  • Molecular weight 357.88 and formula C20H24ClN3O.
  • Soluble in DMSO; available as 10 mM solution or as solid (small pack sizes).
  • Supplied with datasheet, certificate of analysis, and safety data sheet for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.