Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neo 376 (SPI-376) | 496921-73-4 | 99.2% | 357.88 g/mol | C20H24ClN3O | 25 MG
SDP

Catalog No. 5000247226
Encompass_Preferred

NEO 376 (SPI-376) is a small-molecule research compound described as a selective modulator of 5-HT1, GABA, and dopamine receptors with reported antipsychotic activity. Supplied as a white to off-white solid for preclinical research use; available in multiple pack sizes and solution formats.

  • Selective modulator of 5-HT1, GABA, and dopamine receptors.
  • High purity (99.23%) suitable for research applications.
  • Chemical formula C20H24ClN3O; molecular weight 357.88 g/mol.
  • Readily soluble in DMSO; manufacturer provides in vivo formulation protocols.
  • Available as solids (multiple mg sizes) and 10 mM DMSO solutions for convenience.
  • Storage recommendations provided to maintain stability of powder and solutions.

Catalog No. 50-002-47226 Supplier Medchemexpress LLC Supplier No. HY10158325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NEO 376 (SPI-376) is a small-molecule research compound described as a selective modulator of 5-HT1, GABA, and dopamine receptors with reported antipsychotic activity. Supplied as a white to off-white solid for preclinical research use; available in multiple pack sizes and solution formats.

  • Selective modulator of 5-HT1, GABA, and dopamine receptors.
  • High purity (99.23%) suitable for research applications.
  • Chemical formula C20H24ClN3O; molecular weight 357.88 g/mol.
  • Readily soluble in DMSO; manufacturer provides in vivo formulation protocols.
  • Available as solids (multiple mg sizes) and 10 mM DMSO solutions for convenience.
  • Storage recommendations provided to maintain stability of powder and solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.