Learn More
Electron Microscopy Sciences NEO Micro Coverglass, 24 x 40mm, size: 24 x 55mm., 1000/CS
SDP
Catalog No. 502091013
Encompass_Preferred
This Special high-quality flat glass has a workable memory that makes it flexible enough to withstand the stress of automated coverslippers. A special coating privent slips from sticking to each other or reaching to environmental conditions, with no optical interference. Suitable for long-term storage. It is extremely stress resistant and environmentally friendly. size: 24 x 55mm.
Catalog No. 50-209-1013 Supplier Electron Microscopy Sciences Supplier No. 7220730
This Special high-quality flat glass has a workable memory that makes it flexible enough to withstand the stress of automated coverslippers. A special coating privent slips from sticking to each other or reaching to environmental conditions, with no optical interference. Suitable for long-term storage. It is extremely stress resistant and environmentally friendly. size: 24 x 55mm.