Learn More
Electron Microscopy Sciences NEO Micro Coverglass, 24 x 40mm, size: 24 x 60mm., 100/PK
SDP
Catalog No. 502091010
Encompass_Preferred
This Special high-quality flat glass has a workable memory that makes it flexible enough to withstand the stress of automated coverslippers. A special coating privent slips from sticking to each other or reaching to environmental conditions, with no optical interference. Suitable for long-term storage. It is extremely stress resistant and environmentally friendly. size: 24 x 60mm.
Catalog No. 50-209-1010 Supplier Electron Microscopy Sciences Supplier No. 7220704
This Special high-quality flat glass has a workable memory that makes it flexible enough to withstand the stress of automated coverslippers. A special coating privent slips from sticking to each other or reaching to environmental conditions, with no optical interference. Suitable for long-term storage. It is extremely stress resistant and environmentally friendly. size: 24 x 60mm.