Learn More
Med Vet International Neo-Predef Topical Powder 15 Gram
Supplier:  Med Vet International RXNEOPREDEF
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Neo-Predef with Tetracaine Powder is indicated in the treatment or adjunctive therapy of certain ear and skin conditions in dogs, cats and horses caused by or associated with neomycin-susceptible organisms and/or allergy. In addition, it is indicated as superficial dressing applied to minor cuts, wounds, lacerations, abrasions, and for post-surgical application to reduce pain and inflammatory response. May be used as a dusting powder following amputation of tails, claws, and dew-claws; following ear trimming and castrating; and following surgical procedures such as spays. Each gram contains neomycin sulfate, 5 mg (equivalent to 3.5 mg neomycin); isoflupredone acetate, 1 mg; tetracaine hydrochloride, 5 mg; myristyl-gamma-picolinium chloride, 0.2 mg; and lactose hydrous.
Catalog No. 50-282-8355
 Restricted Item