Learn More
TARGETMOL CHEMICALS INC NEO2734 5MG
SDP
Catalog No. 502579056
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NEO2734 (EP31670) is an orally active and selective inhibitor of p300/CBP and BET bromodomain(IC50 of <30 nM for both p300/CBP and BET bromodomains). purity: 98%
Catalog No. 50-257-9056 Supplier TARGETMOL CHEMICALS INC Supplier No. T86585MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NEO2734 (EP31670) is an orally active and selective inhibitor of p300/CBP and BET bromodomain(IC50 of <30 nM for both p300/CBP and BET bromodomains). purity: 98%