Learn More
Echelon Biosciences Research Labs NEOBEADS PIP PURIFICATION SYSTEM
SDP
Catalog No. 50720693
Encompass
The NeoBeads PIP Purification System is useful in biological and chemical applications for purification of lipids. Each milligram of beads will bind at least 1 nmole phosphoinositide. The NeoBeads PIP Purification System allows for selective purification/immobilization of phosphoinositides from biological samples. Phosphoinositides can be recovered from the beads using fresh 2 M aqueous triethylamine bicarbonate, chloroform, methanol (3:2:6; v/v). See the Technical Data Sheet for protocol and further details. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-693 Supplier Echelon Biosciences Research Labs Supplier No. PB99920MG
The NeoBeads PIP Purification System is useful in biological and chemical applications for purification of lipids. Each milligram of beads will bind at least 1 nmole phosphoinositide. The NeoBeads PIP Purification System allows for selective purification/immobilization of phosphoinositides from biological samples. Phosphoinositides can be recovered from the beads using fresh 2 M aqueous triethylamine bicarbonate, chloroform, methanol (3:2:6; v/v). See the Technical Data Sheet for protocol and further details. This item is non returnable