Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neochebulagic acid | 28196-46-5 | 25 MG
SDP

Catalog No. 5000420529
Encompass_Preferred

Neochebulagic acid is isolated from *Phyllanthus emblica L.* and is intended for research use only. This product has been tested and complies with given specifications.

  • Purity: 96.71%
  • Molecular weight: 972.68
  • Molecular formula: C41H32O28
  • Appearance: White to off-white solid
  • Storage: 4°C, protected from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protected from light)
  • Structure classification: Phenols, polyphenols
  • Isolated from *Phyllanthus emblica L.*

Catalog No. 50-004-20529 Supplier Medchemexpress LLC Supplier No. 25MGHYN6909
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Neochebulagic acid is isolated from *Phyllanthus emblica L.* and is intended for research use only. This product has been tested and complies with given specifications.

  • Purity: 96.71%
  • Molecular weight: 972.68
  • Molecular formula: C41H32O28
  • Appearance: White to off-white solid
  • Storage: 4°C, protected from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protected from light)
  • Structure classification: Phenols, polyphenols
  • Isolated from *Phyllanthus emblica L.*

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.