Learn More
Medchemexpress LLC Neochebulagic acid | 28196-46-5 | 50 MG

Supplier: Medchemexpress LLC HYN690950MG
Neochebulagic acid is isolated from Phyllanthus emblica L.
- Purity of 96.71%
- Molecular weight of 972.68
- Chemical formula: C41H32O28
- Appearance: solid
- Color: white to off-white
- Classified as phenols, polyphenols
- Initial source: plants (Combretaceae, Terminalia chebula Retz.)
- Shipping at room temperature in continental US (may vary elsewhere)
- Storage at 4°C, protected from light; in solvent: -80°C for 6 months; -20°C for 1 month (protected from light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.