Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neochlorogenic acid | 906-33-2 | MFCD10566639 | 354.31 | 1 MG
SDP

Catalog No. 5000416965
Encompass_Preferred

Neochlorogenic acid is a natural polyphenolic compound found in dried fruits and other plants. It inhibits the production of TNF-α and IL-1β, suppresses iNOS and COX-2 protein expression, and also inhibits phosphorylated NF-κB p65 and p38 MAPK activation.

  • Inhibits lipopolysaccharide (LPS)-induced NO production by suppressing iNOS and COX-2 protein expression and pro-inflammatory cytokines like TNF-α and IL-1β in BV2 microglia cells.
  • Inhibits phosphorylated p38 MAPK and NF-κB p65 activation in activated microglia.
  • Purity: 99.80%.
  • Appearance: Solid, White to off-white.

Catalog No. 50-004-16965 Supplier Medchemexpress LLC Supplier No. HYN07221MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Neochlorogenic acid is a natural polyphenolic compound found in dried fruits and other plants. It inhibits the production of TNF-α and IL-1β, suppresses iNOS and COX-2 protein expression, and also inhibits phosphorylated NF-κB p65 and p38 MAPK activation.

  • Inhibits lipopolysaccharide (LPS)-induced NO production by suppressing iNOS and COX-2 protein expression and pro-inflammatory cytokines like TNF-α and IL-1β in BV2 microglia cells.
  • Inhibits phosphorylated p38 MAPK and NF-κB p65 activation in activated microglia.
  • Purity: 99.80%.
  • Appearance: Solid, White to off-white.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.