Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Neocuproine hydrate

Catalog No. 1219085G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-190-85G 5 g
12-190-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-190-85G Supplier Sigma Aldrich Supplier No. 1219085G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Neocuproine hydrate is chelating agent and forms complex with cu+ formed by reduction of cu+2 by hydroxylamine hydrochloride and produces electrochemiluminescence
  • It is an efficient ligand for alcohol oxidation with palladium in 1:1 water/dmso mixtures.neocuproine hydrate was used in determination of copper ions in aqueous solution using electrochemiluminescence

Specifications

CAS 654054-57-6
Molecular Formula C14H12N2 -+ xH2O
Linear Formula C14H12N2 -+ xH2O
Quantity 5 g
MDL Number MFCD00149306
Synonym 2,9-Dimethyl-1,10-phenanthroline hydrate
Molecular Weight (g/mol) 208.26 (anhydrous basis)
Percent Purity 99%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.