Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neocuproine Hydrochloride, ≥98% (Dry Basis)

Catalog No. AAJ19430A1 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ19430A1 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ19430A1 Supplier Thermo Scientific Supplier No. J19430A1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

CAS: 7296-20-0 | C14H13ClN2 | 244.722 g/mol

 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7296-20-0
Molecular Formula C14H13ClN2
Molecular Weight (g/mol) 244.722
MDL Number MFCD00150062
InChI Key QUGBNSJLQDNNPK-UHFFFAOYSA-N
Synonym neocuproine hydrochloride, 1,10-phenanthroline, 2,9-dimethyl-, hydrochloride, 1,10-phenanthroline, 2,9-dimethyl-, monohydrochloride, 2,9-dimethyl-1,10-phenanthroline hydrochloride, 2,9-dimethyl-1,10-phenanthroline hcl, 1,10-phenanthroline, 2,9-dimethyl-, hydrochloride 1:?, 1,10-phenanthroline, 2,9-dimethyl-, hydrochloride 1:1, 2,9-dimethylpyridino 3,2-h quinoline, chloride, c14h12n2.clh, neocuproine hcl
PubChem CID 81718
IUPAC Name 2,9-dimethyl-1,10-phenanthroline;hydrochloride
SMILES CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C.Cl

Specifications

Packaging Plastic bottle
Quantity 1 kg
Formula Weight 244.72 (Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Assay Dry basis
Chemical Name or Material Neocuproine hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-732-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.