Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neocuproine hydrochloride monohydrate | 303136-82-5 | 99.15% | 262.73 | 100 MG
SDP

Catalog No. 5000452603
Encompass_Preferred

Neocuproine hydrochloride monohydrate can be used to prepare complex solutions for studying antioxidant capacity determination in biological samples based on copper reduction complexes.

  • Used for research purposes only
  • High purity: 99.15%
  • Available in solid appearance
  • Color: Off-white to light yellow
  • Soluble in DMSO
  • Store at room temperature, keep dry and cool

Catalog No. 50-004-52603 Supplier Medchemexpress LLC Supplier No. HYW011757100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Neocuproine hydrochloride monohydrate can be used to prepare complex solutions for studying antioxidant capacity determination in biological samples based on copper reduction complexes.

  • Used for research purposes only
  • High purity: 99.15%
  • Available in solid appearance
  • Color: Off-white to light yellow
  • Soluble in DMSO
  • Store at room temperature, keep dry and cool

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.