Learn More
Medchemexpress LLC Neocuproine hydrochloride monohydrate | 303136-82-5 | MFCD00150062 | 99.2% | 262.73 | 25 MG
SDP
Catalog No. 5000452126
Encompass_Preferred
Neocuproine hydrochloride monohydrate can be used to prepare complex solutions for studying antioxidant capacity determination in biological samples based on copper reduction complexes.
  • Appearance: solid
  • Color: off-white to light yellow
  • Handling instructions: store at room temperature, keep dry and cool. In solvent: -80°C 1 year, -20°C 6 months.
Catalog No. 50-004-52126 Supplier Medchemexpress LLC Supplier No. 25MGHYW011757
Neocuproine hydrochloride monohydrate can be used to prepare complex solutions for studying antioxidant capacity determination in biological samples based on copper reduction complexes.
  • Appearance: solid
  • Color: off-white to light yellow
  • Handling instructions: store at room temperature, keep dry and cool. In solvent: -80°C 1 year, -20°C 6 months.