Learn More
Miele, Inc. Neodisher EM 5l
Catalog No. 501713107
Encompass_Preferred
Neodisher EM 5lAdditional component for the automated cleaning of laboratory glassware.30 % non-ionic surfactants
Catalog No. 50-171-3107 Supplier Miele, Inc. Supplier No. 94000140
Neodisher EM 5lAdditional component for the automated cleaning of laboratory glassware.30 % non-ionic surfactants