Learn More
Andwin Scientific NEODISHER FLA 10 LTR EA
SDP
Catalog No. NC0527965
Encompass_Preferred
The neodisher Detergents offer consistent, high level cleanliness in Miele glassware washing systems. This product is very useful and efficient in the elimination of organic, inorganic, and metal residues from laboratory glassware and medical and dental instruments in many industries, including cosmetic, wastewater, environmental, biotech, and food. It is most ideal for wastewater, food, environmental, educational, and chemical applications. This product is sold in a quantity of 10 liters.
Catalog No. NC0527965 Supplier Andwin Scientific Supplier No. 62111858
The neodisher Detergents offer consistent, high level cleanliness in Miele glassware washing systems. This product is very useful and efficient in the elimination of organic, inorganic, and metal residues from laboratory glassware and medical and dental instruments in many industries, including cosmetic, wastewater, environmental, biotech, and food. It is most ideal for wastewater, food, environmental, educational, and chemical applications. This product is sold in a quantity of 10 liters.