Learn More
Miele, Inc. Neodisher A8 10kg Powder
Catalog No. 501713102
Encompass_Preferred
Neodisher A8 10kg PowderFor efficient cleaning of blood, protein and food residues.contains 15-30% phosphates, 5% chloine-based bleaching agent
Catalog No. 50-171-3102 Supplier Miele, Inc. Supplier No. 94069003
Neodisher A8 10kg PowderFor efficient cleaning of blood, protein and food residues.contains 15-30% phosphates, 5% chloine-based bleaching agent