Learn More
Miele, Inc. Neodisher F 4x3kg powder
Catalog No. 501713103
Encompass_Preferred
Neodisher F 4x3kg powderFor removal of grease, oil, paraffin, felt pen.Free of phosphates and surfactants, 5-15% polycarboxylates.
This item is not returnable. View return policy
Catalog No. 50-171-3103 Supplier Miele, Inc. Supplier No. 94069004
Neodisher F 4x3kg powderFor removal of grease, oil, paraffin, felt pen.Free of phosphates and surfactants, 5-15% polycarboxylates.