Learn More
Miele, Inc. Neodisher FT 12kg/9l Liquid
Catalog No. 501713112
Encompass_Preferred
Neodisher FT 12kg/9l LiquidFor the automated cleaning of laboratory glassware.With oxidizing effect, free of surfactants
Catalog No. 50-171-3112 Supplier Miele, Inc. Supplier No. 94069014
Neodisher FT 12kg/9l LiquidFor the automated cleaning of laboratory glassware.With oxidizing effect, free of surfactants