Learn More
Medex Supply Neodisher MultiZym 5 liter (1.32 gallons) canister
SDP
Catalog No. NC2822778
Encompass
Excellent cleaning performance due to multi-enzymatic formula. Active residue-free cleaning, prevents redeposition of protein residues. Removes biofilms. Excellent material compatibility, pH-neutral.Detergent for the manual reprocessing of thermostable and thermolabile instrumentsFree of boric acid and boratesFree of perfumes and colourants
Catalog No. NC2822778 Supplier Medex Supply Supplier No. DRW442735L
Excellent cleaning performance due to multi-enzymatic formula. Active residue-free cleaning, prevents redeposition of protein residues. Removes biofilms. Excellent material compatibility, pH-neutral.Detergent for the manual reprocessing of thermostable and thermolabile instrumentsFree of boric acid and boratesFree of perfumes and colourants