Learn More
Andwin Scientific NEODISHER Z 10 LTR EA
SDP
Catalog No. NC0527966
Encompass_Preferred
The neodisher Detergents offer consistent, high level cleanliness in Miele glassware washing systems. This product is very useful and efficient in the elimination of organic, inorganic, and metal residues from laboratory glassware and medical and dental instruments in many industries, including cosmetic, wastewater, environmental, biotech, and food. It is composed only of organic acids. This product is sold in a quantity of 10 liters.
Catalog No. NC0527966 Supplier Andwin Scientific Supplier No. 62111834
The neodisher Detergents offer consistent, high level cleanliness in Miele glassware washing systems. This product is very useful and efficient in the elimination of organic, inorganic, and metal residues from laboratory glassware and medical and dental instruments in many industries, including cosmetic, wastewater, environmental, biotech, and food. It is composed only of organic acids. This product is sold in a quantity of 10 liters.