Learn More
Miele, Inc. Neodisher Z 5 liter
Catalog No. 501713100
Encompass_Preferred
Neodisher Z 5 literNeutralizer for machine cleaning of thermostable and thermolabile instruments and laboratory glassware.Based on organic acids, free of surfactants and phosphates
Catalog No. 50-171-3100 Supplier Miele, Inc. Supplier No. 94069001
Neodisher Z 5 literNeutralizer for machine cleaning of thermostable and thermolabile instruments and laboratory glassware.Based on organic acids, free of surfactants and phosphates