Learn More
Miele, Inc. Neodisher Z 10 liter
Catalog No. 501713101
Encompass_Preferred
Neodisher Z 10 literNeutralizer for machine cleaning of thermostable and thermolabile instruments and laboratory glassware.Based on organic acids, free of surfactants and phosphates
Catalog No. 50-171-3101 Supplier Miele, Inc. Supplier No. 94069002
Neodisher Z 10 literNeutralizer for machine cleaning of thermostable and thermolabile instruments and laboratory glassware.Based on organic acids, free of surfactants and phosphates