Learn More
Strem, An Ascensus Company CAS# 6192-13-8. 50g. Neodymium(III) acetate hydrate (99.9%-Nd) (REO). MFCD00150120
SDP
Catalog No. 5090115964
Encompass
CAS# 6192-13-8. 50g. Neodymium(III) acetate hydrate (99.9%-Nd) (REO). MFCD00150120. Molecular Weight: 321.38. Molecular Formula: Nd(OOCCH3)3∙XH2O. Color/form: light purple xtl. Strem# 93-6020. http://www.strem.com/catalog/v/93-6020/
Catalog No. 50-901-15964 Supplier Strem, An Ascensus Company Supplier No. 93602050G
Add to Cart
Add to Cart
CAS# 6192-13-8. 50g. Neodymium(III) acetate hydrate (99.9%-Nd) (REO). MFCD00150120. Molecular Weight: 321.38. Molecular Formula: Nd(OOCCH3)3∙XH2O. Color/form: light purple xtl. Strem# 93-6020. http://www.strem.com/catalog/v/93-6020/