Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) chloride hydrate, REacton™, 99.9% (REO)

Catalog No. AA1125130
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1125130 250 g
AA1125118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1125130 Supplier Thermo Scientific Chemicals Supplier No. 01125130
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 19423-78-0 | Cl3Nd | 250.59 g/mol

Neodymium(III) chloride hydrate is used for production of neodymium metal. It also finds application in lasers and fiber amplifiers. It serves as dopants for silica based as well as plastic fibers. It plays an important role of additives in organic light emitting diodes resulting in improved LED emission spectrum. In addition, it is used in catalysis of polymerization of various dienes. It is used to modify titanium dioxide for use as a photocatalyst. Its coating on aluminum and its alloy protect them from corrosion. It is also extensively used as fluorescent labels for organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-78-0
Molecular Weight (g/mol) 250.59
MDL Number MFCD00011132
InChI Key ATINCSYRHURBSP-UHFFFAOYSA-K
IUPAC Name trichloroneodymium hexahydrate
SMILES [Cl-].[Cl-].[Cl-].[Nd+3]

Specifications

Melting Point 124°C
Physical Form Crystalline Aggregates
Quantity 250 g
Assay Percent Range 99.9% (REO)
Molecular Formula Cl3Nd
Solubility Information Soluble in water.
Formula Weight 250.60 (Anhydrous)
Odor Odorless
Appearance Crystalline aggregrates
Chemical Name or Material Neodymium(III) chloride hydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-031-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.