Learn More
Strem, An Ascensus Company CAS# 19236-15-8. 1g. Neodymium(III) i-propoxide (99.9%-Nd) (REO). MFCD00058819
SDP
Catalog No. 5090115960
Encompass
CAS# 19236-15-8. 1g. Neodymium(III) i-propoxide (99.9%-Nd) (REO). MFCD00058819. Molecular Weight: 321.51. Molecular Formula: Nd[OCH(CH3)2]3. Color/form: light blue pwdr. Strem# 93-6012. http://www.strem.com/catalog/v/93-6012/
Catalog No. 50-901-15960 Supplier Strem, An Ascensus Company Supplier No. 9360121G
Add to Cart
Add to Cart
CAS# 19236-15-8. 1g. Neodymium(III) i-propoxide (99.9%-Nd) (REO). MFCD00058819. Molecular Weight: 321.51. Molecular Formula: Nd[OCH(CH3)2]3. Color/form: light blue pwdr. Strem# 93-6012. http://www.strem.com/catalog/v/93-6012/