Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Neodymium(III) nitrate hexahydrate

Catalog No. 289175100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-917-5100G 100 g
28-917-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-917-5100G Supplier Sigma Aldrich Supplier No. 289175100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Neodymium(III) nitrate hexahydrate has a variety of uses such as:fabrication of perovskite based solid oxide fuel cells.synthesis of Nd3+ doped vanadium pentoxide nanostructure for potential usage in supercapacitors.a catalyst for Friedlander synthesis of surface modified quinolones for application in medicinal chemistry.

Specifications

CAS 16454-60-7
Quantity 100 g
Molecular Formula N3NdO9 -+ 6H2O
Linear Formula Nd(NO3)3 -+ 6H2O
MDL Number MFCD00149799
Molecular Weight (g/mol) 438.35
Percent Purity 99.9% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.