Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) nitrate hydrate, 99.99% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Alfa Aesar 1124506

Quantity 5g

 View more versions of this product

Catalog No. AA1124506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13746-96-8
99.99% (REO)
330.25 (Anhydrous)
Neodymium(III) nitrate hydrate
Wet Crystalline Aggregates
Nd(NO3)3·xH2O
MFCD00149798

Specifications

13746-96-8
99.99% (REO)
Violet crystals
MFCD00149798
Light blue
Neodymium(III) nitrate hydrate
Wet Crystalline Aggregates
Nd(NO3)3·xH2O
330.25 (Anhydrous)
Odorless
5g

Safety and Handling

SDS & Safety Documents

H272-H315-H319-H335-H303: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 233-153-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit