Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III)-oxide, 99.9%, (trace metal basis)

Catalog No. p-222140
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211580250 25 g Glass bottle
AC211581000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC211580250 Supplier Thermo Scientific Chemicals Supplier No. 211580250
Only null left
Add to Cart

CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-97-9
Molecular Formula Nd2O3
Molecular Weight (g/mol) 336.48
MDL Number MFCD00011134
InChI Key PLDDOISOJJCEMH-UHFFFAOYSA-N
IUPAC Name dineodymium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Nd+3].[Nd+3]

Specifications

Melting Point 2270.0°C
Color Blue
CAS Min % 99.85
CAS Max % 100.0
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.90%
Linear Formula ND2O3
Merck Index 14, 6450
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in acids
Formula Weight 336.47
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 7.24
Density 7.2400g/mL
Chemical Name or Material Neodymium(III)-oxide, 99.9%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 215-214-1
RTECSNumber QP0185000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.