Learn More
Neogen Diagnostics K-Gold PNPP Substrate, 1 L, Microwell, AP, Colorimetric
Supplier: Neogen Diagnostics 303177
K-Gold Substrate is a one-bottle stabilized chromogenic substrate for use with alkaline phosphatase based immunoassays. K-Gold Substrate utilizes the substrate p-nitrophenyl-phosphate (PNPP), which reacts with alkaline phosphatase to produce the chromophore p-nitrophenol. NEOGENs ready-to-use K-Gold formula does not require mixing of additional ingredients or stabilizing agents, making it an ideal formula for kit manufacturers. Stable for a minimum of 30-months when stored at 2-8C.