Learn More
Neogen Diagnostics Progesterone ELISA Kit - 96 well
Catalog No. NC1849084
Encompass_Preferred
Progesterone ELISA is used for the quantitative analysis of Progesterone levels in urine, oral fluid, tissue culture supernatant, plasma or serum samples and is non-species specific. Assay range: 0.04 - 40.0 ng/mL, Assay sample size: 50 µL.
Catalog No. NC1849084 Supplier Neogen Diagnostics Supplier No. 402310
Add to Cart
Add to Cart
Progesterone ELISA is used for the quantitative analysis of Progesterone levels in urine, oral fluid, tissue culture supernatant, plasma or serum samples and is non-species specific. Assay range: 0.04 - 40.0 ng/mL, Assay sample size: 50 µL.