Learn More
Apexbio Technology LLC Neohesperidin 13241-33-3 20mg
SDP
Catalog No. 5000097321
Encompass_Preferred
Neohesperidin is a flavanone glycoside characterized by antioxidant and anti-inflammatory bioactivities It exerts its biological effects primarily by scavenging reactive oxygen species (ROS) and enhancing antioxidant enzyme activities as well as by inhibiting pro-inflammatory mediators including cytokines Investigations employing cell-based assays and animal models utilize Neohesperidin to elucidate protective mechanisms against oxidative damage and inflammation-related tissue injuries Based on these pharmacological properties Neohesperidin holds research potential in the fields of oxidative stress modulation and inflammation-associated disorders
Catalog No. 50-000-97321 Supplier Apexbio Technology LLC Supplier No. N111520
Neohesperidin is a flavanone glycoside characterized by antioxidant and anti-inflammatory bioactivities It exerts its biological effects primarily by scavenging reactive oxygen species (ROS) and enhancing antioxidant enzyme activities as well as by inhibiting pro-inflammatory mediators including cytokines Investigations employing cell-based assays and animal models utilize Neohesperidin to elucidate protective mechanisms against oxidative damage and inflammation-related tissue injuries Based on these pharmacological properties Neohesperidin holds research potential in the fields of oxidative stress modulation and inflammation-associated disorders