Learn More
Medchemexpress LLC Neohesperidin | 13241-33-3 | >98% | 610.56 | 5 G
SDP
Catalog No. 5000427513
Encompass_Preferred
Neohesperidin is a flavonoid compound abundant in citrus plants with antioxidant and anti-inflammatory effects. It can upregulate the Wnt/β-catenin signaling pathway, enhance the nuclear translocation of β-catenin, and promote the differentiation of bone marrow stromal cells.
  • Flavonoid compound
  • Antioxidant effects
  • Anti-inflammatory effects
  • Upregulates Wnt/β-catenin signaling pathway
  • Enhances nuclear translocation of β-catenin
  • Promotes differentiation of bone marrow stromal cells
Catalog No. 50-004-27513 Supplier Medchemexpress LLC Supplier No. HYN01015G
Neohesperidin is a flavonoid compound abundant in citrus plants with antioxidant and anti-inflammatory effects. It can upregulate the Wnt/β-catenin signaling pathway, enhance the nuclear translocation of β-catenin, and promote the differentiation of bone marrow stromal cells.
  • Flavonoid compound
  • Antioxidant effects
  • Anti-inflammatory effects
  • Upregulates Wnt/β-catenin signaling pathway
  • Enhances nuclear translocation of β-catenin
  • Promotes differentiation of bone marrow stromal cells