Learn More
Medchemexpress LLC HY-N0101 50mg Medchemexpress, Neohesperidin CAS:13241-33-3 Purity:>98%
SDP
Catalog No. 501933395
Encompass_Preferred
Medchemexpress, HY-N0101 50mg Neohesperidin CAS:13241-33-3 Neohesperidin is a flavonoid compound found in high amounts in Poncirus trifoliata with anti-oxidant and anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3395 Supplier Medchemexpress LLC Supplier No. HYN010150MG
Medchemexpress, HY-N0101 50mg Neohesperidin CAS:13241-33-3 Neohesperidin is a flavonoid compound found in high amounts in Poncirus trifoliata with anti-oxidant and anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.