Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neohesperidin dihydrochalcone, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013494 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3494 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3494 Supplier MilliporeSigma Supelco Supplier No. 75041100MG
Add to Cart
Add to Cart

Neohesperidin dihydrochalcone is a non-nutritive sweetening agent, widely used as flavor enhancer and flavor modifier in food products. It is also involved in improving the sweetness profile and suppressing bitterness/saltiness properties.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 20702-77-6
Physical Form Neat
Linear Formula C28H36O15
MDL Number MFCD00017711
Melting Point 156°C to 158°C
Synonym 1-(4-((2-O-[6-Deoxy-alpha-L-mannopyranosyl]-beta-D-glucopyranosyl)oxy)-2,6-dihydroxyphenyl)-3-[3-hydroxy-4-methoxyphenyl]-1-propanone; NHDC
Type Analytical Standard
Quantity 100 mg
Formula Weight 612.58
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.