Learn More
Selleck Chemical LLC Neohesperidin 100mg 13241-33-3 NSC 31048
SDP
Supplier:  Selleck Chemical LLC S2332100MG
Encompass_Preferred
Neohesperidin (NSC 31048), a flavanone glycoside found in citrus fruits, is an antioxidant agent. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-726-3