Learn More
Medchemexpress LLC Neohesperidin (Hesperetin 7-O-neohesperidoside) | 13241-33-3 | 98.7% | 610.56 | 500 MG
SDP
Catalog No. 5000427506
Encompass_Preferred
Neohesperidin is a flavonoid compound found in citrus plants, known for its antioxidant and anti-inflammatory properties. It has been shown to upregulate the Wnt/β-catenin signaling pathway and induce apoptosis in human breast adenocarcinoma MDA-MB-231 cells. In vivo, it significantly inhibits HCl/ethanol-induced gastric lesions and improves glucose tolerance and insulin sensitivity in diabetic mice.
  • Possesses antioxidant and anti-inflammatory properties.
  • Upregulates the Wnt/β-catenin signaling pathway.
  • Induces apoptosis in human breast adenocarcinoma MDA-MB-231 cells.
  • Inhibits HCl/ethanol-induced gastric lesions.
  • Improves oral glucose tolerance and insulin sensitivity.
Catalog No. 50-004-27506 Supplier Medchemexpress LLC Supplier No. HYN0101500MG
Neohesperidin is a flavonoid compound found in citrus plants, known for its antioxidant and anti-inflammatory properties. It has been shown to upregulate the Wnt/β-catenin signaling pathway and induce apoptosis in human breast adenocarcinoma MDA-MB-231 cells. In vivo, it significantly inhibits HCl/ethanol-induced gastric lesions and improves glucose tolerance and insulin sensitivity in diabetic mice.
  • Possesses antioxidant and anti-inflammatory properties.
  • Upregulates the Wnt/β-catenin signaling pathway.
  • Induces apoptosis in human breast adenocarcinoma MDA-MB-231 cells.
  • Inhibits HCl/ethanol-induced gastric lesions.
  • Improves oral glucose tolerance and insulin sensitivity.