Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neoline (Bullatine B) | 466-26-2 | MFCD01673484 | 98.0% | 437.57 g/mol | C24H39NO6 | 10 MG
SDP

Catalog No. 5000295523
Encompass_Preferred

Neoline (Bullatine B) is a diterpenoid alkaloid used in preclinical research, reported to have neuroprotective and antinociceptive effects. It is supplied as a solid with high purity and is intended for laboratory applications.

  • Purity 98.0% suitable for research use.
  • Molecular weight 437.57 g/mol; formula C24H39NO6.
  • Available in small milligram quantities for in vitro and in vivo studies.
  • Solid form; store at 4°C and protect from light; in solution, store at -80°C for long term.
  • Synonym: Bullatine B.

Catalog No. 50-002-95523 Supplier Medchemexpress LLC Supplier No. HYN047810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neoline (Bullatine B) is a diterpenoid alkaloid used in preclinical research, reported to have neuroprotective and antinociceptive effects. It is supplied as a solid with high purity and is intended for laboratory applications.

  • Purity 98.0% suitable for research use.
  • Molecular weight 437.57 g/mol; formula C24H39NO6.
  • Available in small milligram quantities for in vitro and in vivo studies.
  • Solid form; store at 4°C and protect from light; in solution, store at -80°C for long term.
  • Synonym: Bullatine B.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.