Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neolinustatin | 72229-42-6 | C17H29NO11 | 5 MG
SDP

Catalog No. 5000375826
Encompass_Preferred

Neolinustatin is a cyanogenic glycoside that can be isolated from flaxseeds.

  • Purity of 99.74%
  • Molecular weight of 423.41
  • Appearance as a solid
  • Color from white to light yellow
  • Classified as other structure
  • Initially sourced from plants Linaceae Linum usitatissimum Linn.
  • Shipping at room temperature in continental US (may vary elsewhere)
  • Storage: 4°C (sealed, away from moisture and light); in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light).

Catalog No. 50-003-75826 Supplier Medchemexpress LLC Supplier No. HY1512455MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neolinustatin is a cyanogenic glycoside that can be isolated from flaxseeds.

  • Purity of 99.74%
  • Molecular weight of 423.41
  • Appearance as a solid
  • Color from white to light yellow
  • Classified as other structure
  • Initially sourced from plants Linaceae Linum usitatissimum Linn.
  • Shipping at room temperature in continental US (may vary elsewhere)
  • Storage: 4°C (sealed, away from moisture and light); in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.