Learn More
Medchemexpress LLC Neoliquiritin | 5088-75-5 | 5 MG
SDP
Catalog No. 5000421683
Encompass_Preferred
Neoliquiritin is a compound isolated from Glycyrrhiza uralensis, known for its anti-inflammatory activity. This white to yellow solid boasts a high purity of 99.85%. It is intended strictly for research purposes and has not been fully validated for medical applications or human consumption.
  • Isolated from Glycyrrhiza uralensis
  • Exhibits anti-inflammatory activity
  • Purity of 99.85%
  • White to yellow solid appearance
  • Suitable for laboratory research
Catalog No. 50-004-21683 Supplier Medchemexpress LLC Supplier No. HYN21235MG
Neoliquiritin is a compound isolated from Glycyrrhiza uralensis, known for its anti-inflammatory activity. This white to yellow solid boasts a high purity of 99.85%. It is intended strictly for research purposes and has not been fully validated for medical applications or human consumption.
  • Isolated from Glycyrrhiza uralensis
  • Exhibits anti-inflammatory activity
  • Purity of 99.85%
  • White to yellow solid appearance
  • Suitable for laboratory research